2-Bromoterephthalic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14393-06
A14393-14
A14393-22
CAAAA14393-06EA
135.56
CAD
CAAAA14393-06
CAAAA14393-14
CAAAA14393-22
2-Bromoterephthalic acid 97%
2-Bromoterephthalic acid
Purity: 97%
CAS No.: 586-35-6
Molecular Formula: C8H5BrO4
Formula Weight: 245.03
Melting Point: 297-300°
Beilstein Reference: 2616163
UN No.: 2811
MDL No.: MFCD00002403
Formula:
C₈H₅BrO₄ Melting Pt: 297…300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002403 CAS Number: 586-35-6 EINECS: 209-572-8 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...