6-Azauracil 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2,4-Triazine-3,5(2H,4H)-dione

A14389-06 A14389-14
CAAAA14389-06EA 95.59 CAD
CAAAA14389-06 CAAAA14389-14
6-Azauracil 98%
6-Azauracil

Purity: 98% CAS No.: 461-89-2 Molecular Formula: C3H3N3O2 Formula Weight: 113.08 Melting Point: 274-282° Beilstein Reference: 116472

Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
Melting Pt: 274…282 °C
Storage Temperature: Ambient
MDL Number: MFCD00006456
CAS Number: 461-89-2
EINECS: 207-318-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR