Cysteaminium chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14377-14
A14377-22
A14377-36
CAAAA14377-14EA
93.22
CAD
CAAAA14377-14
CAAAA14377-22
CAAAA14377-36
Cysteaminium chloride 98+%
Cysteaminium chloride
An inhibitor of DMBA-induced mammary tumors.
Purity: 98+%
CAS No.: 156-57-0
Molecular Formula: HSCH2CH2NH2.HCl
Formula Weight: 113.61
Melting Point: 65-69°
Solubility: Soluble in water and alcohol
Beilstein Reference: 3590083
Merck Reference: 14,2779
MDL No.: MFCD00012904
Soluble in water and alcohol
Formula:
C₂H₇NS·HCl Melting Pt: 65…69 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00012904 CAS Number: 156-57-0 EINECS: 205-858-1 UN: 3335 ADR: 9,III Merck Index: 14,2779 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...