Methyl-3-(trifluoromethyl)benzoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14316-06
A14316-14
CAAAA14316-06EA
74.73
CAD
CAAAA14316-06
CAAAA14316-14
Methyl-3-(trifluoromethyl)benzoate 97%
Methyl 3-(trifluoromethyl)benzoate
Purity: 97%
CAS No.: 2557-13-3
Molecular Formula: C9H7F3O2
Formula Weight: 204.15
Flash Point: 84°(183°F)
Refractive Index: 1.4530
Boiling Point: 80-82°/15mm
Beilstein Reference: 1963428
MDL No.: MFCD00043543
Formula:
C₉H₇F₃O₂ Boiling Pt: 80…82 °C (15 mmHg) Density: 1.295 Flash Pt: 84 °C (183 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00043543 CAS Number: 2557-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...