(p-Toluenesulfonyl)methyl isocyanide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14312-06
A14312-14
A14312-22
CAAAA14312-06EA
60.04
CAD
CAAAA14312-06
CAAAA14312-14
CAAAA14312-22
(p-Toluenesulfonyl)methyl isocyanide 97%
(p-Toluenesulphonyl)methyl isocyanide
Fieser: 4,514 5,684 6,600 7,377 8,493 10,409 12,511 13,313
Purity: 97+%
CAS No.: 36635-61-7
Molecular Formula: C9H9NO2S
Formula Weight: 195.24
Melting Point: 109-114°
Beilstein Reference: 3592382
UN No.: 2811
MDL No.: MFCD00000005
Formula:
C₉H₉NO₂S Melting Pt: 109…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000005 CAS Number: 36635-61-7 EINECS: 253-140-1 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...