Perfluoro(methylcyclohexane) 94%
Supplier: Thermo Scientific Chemicals
Synonyms:
FlutecR PP2
A14233-22
A14233-14
CAAAA14233-22EA
413.96
CAD
CAAAA14233-22
CAAAA14233-36
CAAAA14233-14
Perfluoro(methylcyclohexane) 94%
Perfluoro(methylcyclohexane)
Purity: 94%
CAS No.: 355-02-2
Molecular Formula: C7F14
Formula Weight: 350.06
Refractive Index: 1.3 00
Odor: Odorless
Boiling Point: 75-76°
Beilstein Reference: 1915981
MDL No.: MFCD00001468
Formula:
C₇F₁₄ Boiling Pt: 75…76 °C Melting Pt: –70 °C Density: 1,800 Storage Temperature: Ambient |
MDL Number:
MFCD00001468 CAS Number: 355-02-2 EINECS: 206-573-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...