Triethyl phosphonoacetate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl 2-(diethoxyphosphoryl)acetate, Diethyl [(ethoxycarbonyl)methyl]phosphonate

A14120-0E A14120-22 A14120-36
CAAAA14120-0EEA 1979.74 CAD
CAAAA14120-0E CAAAA14120-22 CAAAA14120-36
Triethyl phosphonoacetate 98+%
Triethyl phosphonoacetate

Fieser: 1,1216 6,612 12,527 13,322


Purity: 98+% CAS No.: 867-13-0 Molecular Formula: C8H17O5P Formula Weight: 224.19 Flash Point: 165°(329°F) Refractive Index: 1.4310 Melting Point: -24° Boiling Point: 260-262° Beilstein Reference: 1343714 MDL No.: MFCD00009177

Formula: C₈H₁₇O₅P
MW: 224.19 g/mol
Boiling Pt: 260…262 °C
Melting Pt: –24 °C
Density: 1.125
Flash Pt: 165 °C (329 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009177
CAS Number: 867-13-0
EINECS: 212-757-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR