Methyl-2-furoate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methyl 2-Furancarboxylate, Methyl furan-2-carboxylate, Methyl 2-furoate
A14009-14
A14009-22
A14009-36
CAAAA14009-14EA
47.72
CAD
CAAAA14009-14
CAAAA14009-22
CAAAA14009-36
Methyl-2-furoate 98+%
Methyl 2-furoate
Purity: 98+%
CAS No.: 611-13-2
Molecular Formula: C6H6O3
Formula Weight: 126.11
Flash Point: 73°(163°F)
Refractive Index: 1.4870
Boiling Point: 180-181°
Beilstein Reference: 111110
Merck Reference: 14,4307
UN No.: 2810
MDL No.: MFCD00003236
Formula:
C₆H₆O₃ Boiling Pt: 180…181 °C Density: 1.179 Flash Pt: 73 °C(163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003236 CAS Number: 611-13-2 EINECS: 210-254-6 UN: 2810 ADR: "6.1",III Merck Index: 14,4307 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...