Ethyl picolinate 99%
Supplier: Thermo Scientific Chemicals
A13973-14
A13973-22
A13973-30
A13973-18
CAAAAA13973-14EA
125.14
CAD
CAAAAA13973-14
CAAAAA13973-22
CAAAAA13973-30
CAAAA13973-18
Ethyl picolinate 99%
Ethyl picolinate
Purity: 99%
CAS No.: 2524-52-9
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Flash Point: 107°(224°F)
Refractive Index: 1.5105
Odor: Characteristic
Melting Point: 1-3°
Boiling Point: 239-241°
Beilstein Reference: 122485
MDL No.: MFCD00006292
Formula:
C₈H₉NO₂ Boiling Pt: 239…241 °C Melting Pt: 1…3 °C Density: 1.120 Flash Pt: 107 °C(224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006292 CAS Number: 2524-52-9 EINECS: 219-758-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...