3-Methylthiophene-2-carboxaldehyde tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13961-14
A13961-22
CAAAA13961-14EA
65.41
CAD
CAAAA13961-14
CAAAA13961-22
3-Methylthiophene-2-carboxaldehyde tech. 90%, Technical Grade
3-Methylthiophene-2-carboxaldehyde
Purity: 90%
CAS No.: 5834-16-2
Molecular Formula: C6H6OS
Formula Weight: 126.18
Flash Point: 82°(179°F)
Refractive Index: 1.5860
Boiling Point: 215°
Beilstein Reference: 107874
MDL No.: MFCD00005430
Formula:
C₆H₆OS Boiling Pt: 215 °C Density: 1.170 Flash Pt: 82 °C(179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005430 CAS Number: 5834-16-2 EINECS: 227-418-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...