Ethyl-2-furoate 99%
Supplier: Thermo Scientific Chemicals
A13909-14
A13909-22
A13909-36
CAAAA13909-14EA
43.92
CAD
CAAAA13909-14
CAAAA13909-22
CAAAA13909-36
Ethyl-2-furoate 99%
Ethyl 2-furoate
Purity: 99%
CAS No.: 614-99-3
Molecular Formula: C7H8O3
Formula Weight: 140.14
Storage Temperature: 2°C to 8°C
Flash Point: 70°(158°F)
Melting Point: 31-36°
Boiling Point: 195-196°
Beilstein Reference: 2653
Merck Reference: 14,4307
MDL No.: MFCD00003237
Formula:
C₇H₈O₃ Boiling Pt: 195…196 °C Melting Pt: 31…36 °C Density: 1.120 Flash Pt: 70 °C(158 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003237 CAS Number: 614-99-3 EINECS: 210-404-0 Merck Index: 14,4307 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...