1,2-Phenylenediacetic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13851-06
A13851-14
CAAAA13851-06EA
32.55
CAD
CAAAA13851-06
CAAAA13851-14
1,2-Phenylenediacetic acid 98+%
1,2-Phenylenediacetic acid
Purity: 98+%
CAS No.: 7500-53-0
Molecular Formula: C10H10O4
Formula Weight: 194.19
Odor: Odorless
Melting Point: 150-152°
Beilstein Reference: 2616747
MDL No.: MFCD00004329
Formula:
C₁₀H₁₀O₄ Melting Pt: 150…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004329 CAS Number: 7500-53-0 EINECS: 231-355-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...