N,N'-Diphenylbenzidine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
4,4'-Bis(phenylamino)biphenyl, N4,N4'-Diphenyl-[1,1'-biphenyl]-4,4'-diamine
A13802-06
A13802-14
CAAAA13802-06EA
100.01
CAD
CAAAA13802-06
CAAAA13802-14
N,N'-Diphenylbenzidine 98%
N,N'-Diphenylbenzidine
Purity: 98%
CAS No.: 531-91-9
Molecular Formula: C24H20N2
Formula Weight: 336.44
Odor: Odorless
Melting Point: 246-250°
Beilstein Reference: 2220993
Merck Reference: 14,3319
MDL No.: MFCD00003016
Formula:
C₂₄H₂₀N₂ Melting Pt: 246…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003016 CAS Number: 531-91-9 EINECS: 208-521-7 Merck Index: 14,3319 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...