Phenylphosphonic dichloride 90+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13777-0B
A13777-18
A13777-30
CAAAA13777-0BEA
690.46
CAD
CAAAA13777-0B
CAAAA13777-18
CAAAA13777-30
Phenylphosphonic dichloride 90+%
Phenylphosphonic dichloride
Purity: 90+%
CAS No.: 824-72-6
Molecular Formula: C6H5Cl2OP
Formula Weight: 194.99
Flash Point: 204°(399°F)
Refractive Index: 1.5600
Melting Point: 1-3°
Boiling Point: 258°
Beilstein Reference: 1072240
UN No.: 3265
MDL No.: MFCD00002070
Formula:
C₆H₅Cl₂OP Boiling Pt: 258 °C Melting Pt: 1…3 °C Density: 1.379 Flash Pt: 204 °C(399 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002070 CAS Number: 824-72-6 EINECS: 212-534-3 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...