4-(Allyloxy)benzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

A13773-09 A13773-18
CAAAA13773-09EA 85.95 CAD
CAAAA13773-09 CAAAA13773-18 CAAAA13773-30
4-(Allyloxy)benzaldehyde 97%
4-(Allyloxy)benzaldehyde

Purity: 97% CAS No.: 40663-68-1 Molecular Formula: C10H10O2 Formula Weight: 162.19 Flash Point: 110°(230°F) Refractive Index: 1.5695 Boiling Point: 150-152°/18mm Beilstein Reference: 2206924 MDL No.: MFCD00014133

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 150…152 °C (18 mmHg)
Density: 1.058
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014133
CAS Number: 40663-68-1
EINECS: 255-027-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR