Acetoin monomer + dimer 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13752-0B
A13752-18
A13752-30
CAAAA13752-0BEA
829.5
CAD
CAAAA13752-0B
CAAAA13752-18
CAAAA13752-30
Acetoin monomer + dimer 95%
Acetoin
Purity: 95%
CAS No.: 513-86-0
Molecular Formula: C4H8O2
Formula Weight: 88.11
Storage Temperature: 2°C to 8°C
Flash Point: 41°(106°F)
Odor: Characteristic
Boiling Point: 148°
Beilstein Reference: 385636
Merck Reference: 14,64
UN No.: 2621
MDL No.: MFCD00038696
Formula:
C₄H₈O₂ Boiling Pt: 148 °C Melting Pt: 20 °C Density: 1.013 Flash Pt: 41 °C (106 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00038696 CAS Number: 513-86-0 EINECS: 208-174-1 UN: 2621 ADR: 3,III Merck Index: 14,64 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...