Diethyl (iodomethyl)phosphonate 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13697-06 A13697-14 A13697-22
CAAAA13697-06EA 153.1 CAD
CAAAA13697-06 CAAAA13697-14 CAAAA13697-22
Diethyl (iodomethyl)phosphonate 98+%
Diethyl (iodomethyl)phosphonate

Purity: 98+% CAS No.: 10419-77-9 Molecular Formula: C5H12IO3P Formula Weight: 278.03 Flash Point: 110°(230°F) Refractive Index: 1.4995 Boiling Point: 112-114°/1mm Beilstein Reference: 1812052 MDL No.: MFCD00041386

Formula: C₅H₁₂IO₃P
Boiling Pt: 112…114 °C/1 mm
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041386
CAS Number: 10419-77-9
EINECS: 233-895-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR