2-Furohydrazide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13630-09
A13630-18
CAAAAA13630-30EA
1980.88
CAD
CAAAAA13630-30
CAAAA13630-09
CAAAA13630-18
2-Furohydrazide 98%
2-Furohydrazide
Purity: 98%
CAS No.: 3326-71-4
Molecular Formula: C5H6N2O2
Formula Weight: 126.12
Melting Point: 77-81°
Beilstein Reference: 114435
Formula:
C₅H₆N₂O₂ Melting Pt: 77…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003235 CAS Number: 3326-71-4 EINECS: 222-046-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...