Phenyl phosphorodiamidate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13562-09
A13562-18
CAAAA13562-09EA
620.94
CAD
CAAAA13562-09
CAAAA13562-18
CAAAA13562-30
Phenyl phosphorodiamidate 98+%
Phenyl phosphorodiamidate
Fieser: 5,517
Purity: 98+%
CAS No.: 7450-69-3
Molecular Formula: C6H9N2O2P
Formula Weight: 172.12
Melting Point: 180-185°
Beilstein Reference: 608629
Formula:
C₆H₉N₂O₂P Melting Pt: 180…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014767 CAS Number: 7450-69-3 EINECS: 231-218-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...