Dioctyl phenylphosphonate 97%

Supplier: Thermo Scientific Chemicals

A13468-06 A13468-14 A13468-22
CAAAAA13468-06EA 85.32 CAD
CAAAAA13468-06 CAAAAA13468-14 CAAAAA13468-22
Dioctyl phenylphosphonate 97%
Dioctyl phenylphosphonate

Purity: 97% CAS No.: 1754-47-8 Molecular Formula: C22H39O3P Formula Weight: 382.53 Flash Point: 110°(230°F) Refractive Index: 1.4795 Boiling Point: 219-220°/0.7mm Beilstein Reference: 2948496 MDL No.: MFCD00009561

Formula: C₂₂H₃₉O₃P
Boiling Pt: 219…220 °C/0.7 mm
Density: 0,932
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009561
CAS Number: 1754-47-8
EINECS: 217-142-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR