trans,trans-2,4-Undecadienal 90+% remainder mainly trans, cis isomer
Supplier: Thermo Scientific Chemicals
A13419-06
A13419-14
CAAAA13419-06EA
39.18
CAD
CAAAA13419-06
CAAAA13419-14
trans,trans-2,4-Undecadienal 90+% remainder mainly trans, cis isomer
trans,trans-2,4-Undecadienal
Purity: 90+%
CAS No.: 30361-29-6
Molecular Formula: C11H18O
Formula Weight: 166.27
Storage Temperature: 2°C to 8°C
Flash Point: 107°(225°F)
Refractive Index: 1.5120
Boiling Point: 128-129°/10mm
Beilstein Reference: 1706330
MDL No.: MFCD00014677
Formula:
C₁₁H₁₈O Boiling Pt: 128…129 °C/10 mm Density: 0,870 Flash Pt: 107 °C(225 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00014677 CAS Number: 30361-29-6 EINECS: 250-148-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...