5-Bromothiophene-2-carbaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromo-2-thiophenecarboxaldehyde

A13381-06 A13381-14 A13381-22
CAAAA13381-06EA 55.14 CAD
CAAAA13381-06 CAAAA13381-14 CAAAA13381-22
5-Bromothiophene-2-carbaldehyde 97%
5-Bromothiophene-2-carbaldehyde

Purity: 97% CAS No.: 4701-17-1 Molecular Formula: C5H3BrOS Formula Weight: 191.05 Storage Temperature: 2°C to 8°C Flash Point: 98°(208°F) Refractive Index: 1.6390 Boiling Point: 104-106°/10mm Beilstein Reference: 108404 MDL No.: MFCD00005432

Formula: C₅H₃BrOS
MW: 191.05 g/mol
Boiling Pt: 104…106 °C/10 mm
Density: 1.762
Flash Pt: 98 °C(208 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005432
CAS Number: 4701-17-1
EINECS: 225-176-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR