3-Quinuclidinone hydrochloride 98+%
Supplier: Thermo Scientific Chemicals
A13320-14
A13320-22
CAAAA13320-06EA
40.61
CAD
CAAAA13320-06
CAAAA13320-14
CAAAA13320-22
3-Quinuclidinone hydrochloride 98+%
3-Quinuclidinone hydrochloride
Purity: 98+%
CAS No.: 1193-65-3
Molecular Formula: C7H11NO.HCl
Formula Weight: 161.63
Melting Point: ca 300° dec.
Beilstein Reference: 3695039
Formula:
C₇H₁₂ClNO Melting Pt: ca 300 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00137391 CAS Number: 1193-65-3 EINECS: 214-776-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...