4-Bromothiophene-2-carbaldehyde 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-2-thiophenecarboxaldehyde, 4-Bromothiophene-2-carboxaldehyde

A13304-06 A13304-14 A13304-22
CAAAA13304-06EA 47.24 CAD
CAAAA13304-06 CAAAA13304-14 CAAAA13304-22
4-Bromothiophene-2-carbaldehyde 96%
4-Bromothiophene-2-carbaldehyde

Purity: 96% CAS No.: 18791-75-8 Molecular Formula: C5H3BrOS Formula Weight: 191.05 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 44-48° Boiling Point: 114-115°/11mm MDL No.: MFCD00005431

Formula: C₅H₃BrOS
MW: 191.05 g/mol
Boiling Pt: 114…115 °C (11 mmHg)
Melting Pt: 44…48 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005431
CAS Number: 18791-75-8
EINECS: 242-577-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR