1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,4-Dihydronaphthalen-1(2H)-one
A13303-22
A13303-36
CAAAA13303-22EA
92.59
CAD
CAAAA13303-22
CAAAA13303-36
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) 97%
1,2,3,4-Tetrahydronaphthalen-1-one (1-Tetralone, α-Tetralone)
Purity: 97%
CAS No.: 529-34-0
Molecular Formula: C10H10O
Formula Weight: 146.19
Flash Point: 129°(264°F)
Refractive Index: 1.5695
Melting Point: 6-8°
Boiling Point: 257°
Beilstein Reference: 607374
MDL No.: MFCD00001688
Formula:
C₁₀H₁₀O MW: 146.19 g/mol Boiling Pt: 257 °C Melting Pt: 6…8 °C Density: 1.097 Flash Pt: 129 °C (264 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001688 CAS Number: 529-34-0 EINECS: 208-460-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...