trans,trans-2,4-Hexadienal predominantly trans 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13200-06
A13200-14
A13200-22
CAAAA13200-06EA
42.5
CAD
CAAAA13200-06
CAAAA13200-14
CAAAA13200-22
trans,trans-2,4-Hexadienal predominantly trans 95%
trans,trans-2,4-Hexadienal
Purity: 95%
CAS No.: 142-83-6
Molecular Formula: C6H8O
Formula Weight: 96.13
Storage Temperature: 2°C to 8°C
Flash Point: 67°(152°F)
Refractive Index: 1.5386
Odor: Floral
Boiling Point: 170-173°
Beilstein Reference: 1698401
UN No.: 2922
MDL No.: MFCD00007004
Formula:
C₆H₈O Boiling Pt: 170…173 °C Density: 0.871 Flash Pt: 67 °C(152 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007004 CAS Number: 142-83-6 EINECS: 205-564-3 UN: 2810 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...