o-Phenylenediacetonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Phenylenediacetonitrile, 1,2-Bis(cyanomethyl)-benzene

A13142-18 A13142-06
CAAAAA13142-18EA 274.92 CAD
CAAAAA13142-18 CAAAA13142-06
o-Phenylenediacetonitrile 98%
o-Phenylenediacetonitrile

Purity: 98% CAS No.: 613-73-0 Molecular Formula: C10H8N2 Formula Weight: 156.19 Melting Point: 57-61° Beilstein Reference: 510075 UN No.: 3439 MDL No.: MFCD00001905

Formula: C₁₀H₈N₂
MW: 156.19 g/mol
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00001905
CAS Number: 613-73-0
EINECS: 210-351-3
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR