4-(Dimethylamino)benzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13117-06
A13117-14
CAAAA13117-06EA
60.67
CAD
CAAAA13117-06
CAAAA13117-14
4-(Dimethylamino)benzonitrile 98%
4-(Dimethylamino)benzonitrile
Purity: 98%
CAS No.: 1197-19-9
Molecular Formula: C9H10N2
Formula Weight: 146.19
Melting Point: 72-76°
Boiling Point: 317-319°
UN No.: 3439
MDL No.: MFCD00001815
Formula:
C₉H₁₀N₂ Boiling Pt: 317…319 °C Melting Pt: 72…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001815 CAS Number: 1197-19-9 EINECS: 214-819-8 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...