2,6-Di-tert-butyl-p-benzoquinone 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,6-Di-tert-butylcyclohexa-2,5-diene-1,4-dione

A13091-06 A13091-14
CAAAA13091-06EA 125.29 CAD
CAAAA13091-06 CAAAA13091-14 CAAAA13091-22
2,6-Di-tert-butyl-p-benzoquinone 98+%
2,6-Di-tert-butyl-p-benzoquinone

Fieser: 9,139


Purity: 98+% CAS No.: 719-22-2 Molecular Formula: C14H20O2 Formula Weight: 220.31 Flash Point: 110°(230°F) Melting Point: 65-69° Beilstein Reference: 1911512 MDL No.: MFCD00001601

Formula: C₁₄H₂₀O₂
Melting Pt: 65…69 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001601
CAS Number: 719-22-2
EINECS: 211-946-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR