Aluminium tri-sec-butanolate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13044-0E
A13044-22
A13044-36
CAAAA13044-0EEA
775.78
CAD
CAAAA13044-0E
CAAAA13044-22
CAAAA13044-36
Aluminium tri-sec-butanolate 97%
Aluminium tri-sec-butanolate
Purity: 97%
CAS No.: 2269-22-9
Molecular Formula: Al[OCH(CH3)CH2CH3]3
Formula Weight: 246.32
Flash Point: 27°(82°F)
Refractive Index: 1.4390
Odor: Alcohol-like
Boiling Point: 200-206°/30mm
Beilstein Reference: 3910908
UN No.: 1992
MDL No.: MFCD00009327
Formula:
C₁₂H₂₇AlO₃ Boiling Pt: 200…206 °C/30 mm Density: 0.967 Flash Pt: 27 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009327 CAS Number: 2269-22-9 EINECS: 218-871-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...