3,4-(Methylenedioxy)aniline 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12975-09
A12975-18
A12975-30
CAAAA12975-09EA
60.83
CAD
CAAAA12975-09
CAAAA12975-18
CAAAA12975-30
3,4-(Methylenedioxy)aniline 98+%
3,4-(Methylenedioxy)aniline
Purity: 98+%
CAS No.: 14268-66-7
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Flash Point: 142°(287°F)
Melting Point: 40-45°
Boiling Point: 144°/16mm
Beilstein Reference: 4919
UN No.: 2811
MDL No.: MFCD00005832
Formula:
C₇H₇NO₂ Boiling Pt: 144 °C/16 mm Melting Pt: 40…45 °C Flash Pt: 142 °C(287 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005832 CAS Number: 14268-66-7 EINECS: 238-161-6 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...