Isopropyltriphenylphosphonium iodide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12881-22
A12881-09
A12881-30
CAAAAA12881-22EA
426.6
CAD
CAAAAA12881-22
CAAAA12881-09
CAAAA12881-30
Isopropyltriphenylphosphonium iodide 98+%
Isopropyltriphenylphosphonium iodide
Purity: 98+%
CAS No.: 24470-78-8
Molecular Formula: (CH3)2CHP(C6H5)3I
Formula Weight: 432.29
Melting Point: 195-198°
Beilstein Reference: 3599629
Formula:
C₂₁H₂₂IP Melting Pt: 195…198 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031595 CAS Number: 24470-78-8 EINECS: 246-281-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...