(Phenylthio)acetonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12869-14
A12869-22
CAAAA12869-06EA
153.48
CAD
CAAAA12869-06
CAAAA12869-14
CAAAA12869-22
(Phenylthio)acetonitrile 98%
(Phenylthio)acetonitrile
Purity: 98%
CAS No.: 5219-61-4
Molecular Formula: C6H5SCH2CN
Formula Weight: 149.22
Flash Point:
Formula:
C₈H₇NS Boiling Pt: 146…147 °C/14 mm Density: 1.142 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00019831 CAS Number: 5219-61-4 EINECS: 226-013-3 UN: 3276 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...