4-Picolyl chloride hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12859-06
A12859-14
A12859-22
CAAAA12859-06EA
54.19
CAD
CAAAA12859-06
CAAAA12859-14
CAAAA12859-22
4-Picolyl chloride hydrochloride 98%
4-Picolyl chloride hydrochloride
Fieser: 4,391
Purity: 98%
CAS No.: 1822-51-1
Molecular Formula: C6H62N.HCl
Formula Weight: 164.04
Melting Point: 166-171°
Beilstein Reference: 3689154
UN No.: 3261
MDL No.: MFCD00012826
Formula:
C₆H₇Cl₂N Melting Pt: 166…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012826 CAS Number: 1822-51-1 EINECS: 217-350-7 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...