2-(4-Chlorophenylthio)benzaldehyde 98%
Supplier: Thermo Scientific Chemicals
A12852-06
A12852-14
CAAAA12852-06EA
125.45
CAD
CAAAA12852-06
CAAAA12852-14
2-(4-Chlorophenylthio)benzaldehyde 98%
2-(4-Chlorophenylthio)benzaldehyde
Purity: 98%
CAS No.: 107572-07-6
Molecular Formula: C13H9ClOS
Formula Weight: 248.72
Melting Point: 71-74°
Boiling Point: 102-104°/14mm
MDL No.: MFCD00051766
Formula:
C₁₃H₉ClOS MW: 248.73 g/mol Boiling Pt: 102…104 °C/14 mm Melting Pt: 71…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051766 CAS Number: 107572-07-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...