Methoxyhydroquinone 97%

Supplier: Thermo Scientific Chemicals
Warning

A12847-18 A12847-30
CAAAA12847-18EA 290.72 CAD
CAAAA12847-18 CAAAA12847-30
Methoxyhydroquinone 97%
Methoxyhydroquinone

Purity: 97% CAS No.: 824-46-4 Molecular Formula: C7H8O3 Formula Weight: 140.14 Melting Point: 87-91° Beilstein Reference: 2045285

Formula: C₇H₈O₃
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00013971
CAS Number: 824-46-4
EINECS: 212-530-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR