2-Methyl-1,3-dioxolane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12824-09
A12824-18
CAAAAA12824-09EA
84.37
CAD
CAAAAA12824-09
CAAAAA12824-18
CAAAAA12824-30
2-Methyl-1,3-dioxolane 98%
2-Methyl-1,3-dioxolane
Purity: 98%
CAS No.: 497-26-7
Molecular Formula: C4H8O2
Formula Weight: 88.11
Flash Point: -2°(28°F)
Refractive Index: 1.3980
Boiling Point: 80-83°
Beilstein Reference: 102520
UN No.: 1993
MDL No.: MFCD00003210
Formula:
C₄H₈O₂ Boiling Pt: 80…83 °C Density: 0.982 Flash Pt: -2 °C(28 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003210 CAS Number: 497-26-7 EINECS: 207-841-4 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...