Methyl-3-aminothiophene-2-carboxylate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12802-06
A12802-14
A12802-22
CAAAA12802-06EA
63.04
CAD
CAAAA12802-06
CAAAA12802-14
CAAAA12802-22
Methyl-3-aminothiophene-2-carboxylate 98+%
Methyl 3-aminothiophene-2-carboxylate
Purity: 98+%
CAS No.: 22288-78-4
Molecular Formula: C6H7NO2S
Formula Weight: 157.19
Melting Point: 63-67°
Boiling Point: 100-102°/0.1mm
MDL No.: MFCD00009765
Formula:
C₆H₇NO₂S Boiling Pt: 100…102 °C/0.1 mm Melting Pt: 63…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009765 CAS Number: 22288-78-4 EINECS: 244-895-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...