tert-Butylchlorodiphenylsilane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
tert-Butyldiphenylchlorosilane, tert-Butyl-diphenylchlorosilane
A12721-09
A12721-18
CAAAA12721-09EA
83.74
CAD
CAAAA12721-09
CAAAA12721-18
tert-Butylchlorodiphenylsilane 97%
tert-Butylchlorodiphenylsilane
Fieser: 6,81 14,60 15,55 18,71 21,75
Purity: 97%
CAS No.: 58479-61-1
Molecular Formula: C16H19ClSi
Formula Weight: 274.87
Flash Point: 112°(233°F)
Physical Form: Liquid
Density: 1.072
Refractive Index: 1.5680
Sensitivity: Moisture Sensitive
Odor: Pungent
Boiling Point: 90-92°/0.01mm
Beilstein Reference: 644023
UN No.: 2987
MDL No.: MFCD00000497
Formula:
C₁₆H₁₉ClSi MW: 274.87 g/mol Boiling Pt: 90…92 °C/0.01 mm Density: 1.072 Flash Pt: 112 °C(233 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000497 CAS Number: 58479-61-1 EINECS: 261-282-0 UN: 2987 ADR: "8",II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...