Pyromellitic dianhydride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2,4,5-Benzenetetracarboxylic anhydride

A12712-0E A12712-22 A12712-36
CAAAA12712-0EEA 1031.74 CAD
CAAAA12712-0E CAAAA12712-22 CAAAA12712-36
Pyromellitic dianhydride 97%
Pyromellitic dianhydride

Purity: 97% CAS No.: 89-32-7 Molecular Formula: C10H2O6 Formula Weight: 218.12 Flash Point: 380°(716°F) Melting Point: 284-288° Boiling Point: 305-310°/30mm Beilstein Reference: 213583 MDL No.: MFCD00005005

Formula: C₁₀H₂O₆
MW: 218.12 g/mol
Boiling Pt: 305…310 °C (30 mmHg)
Melting Pt: 284…288 °C
Density: 1.68
Flash Pt: 380 °C (716 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005005
CAS Number: 89-32-7
EINECS: 201-898-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR