3-Acetylcoumarin 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Acetyl-2H-chromen-2-one

A12532-06 A12532-14
CAAAA12532-06EA 78.37 CAD
CAAAA12532-06 CAAAA12532-14
3-Acetylcoumarin 98+%
3-Acetylcoumarin

Purity: 98+% CAS No.: 3949-36-8 Molecular Formula: C11H8O3 Formula Weight: 188.18 Melting Point: 119-124° Beilstein Reference: 143498

Formula: C₁₁H₈O₃
MW: 188.18
Melting Pt: 119…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00006853
CAS Number: 3949-36-8
EINECS: 223-541-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR