4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) 98+% free radical
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2,6,6-Tetramethyl-4-hydroxy-1-piperidinyloxy radical, 4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, Tempol
A12497-03
A12497-06
A12497-14
CAAAA12497-03EA
57.04
CAD
CAAAA12497-03
CAAAA12497-06
CAAAA12497-14
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) 98+% free radical
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-Hydroxy-TEMPO, TEMPOL, free radical)
Superoxide scavenger with neuroprotective, anti-inflammatory and analgesic effects
Purity: 98+%
CAS No.: 2226-96-2
Molecular Formula: C9H18NO2
Formula Weight: 172.25
Storage Temperature: 2°C to 8°C
Flash Point: 146°(295°F)
pH: 8.2
Melting Point: 69-72°
Beilstein Reference: 1422990
MDL No.: MFCD00006478
Formula:
C₉H₁₈NO₂ Melting Pt: 69…72 °C Flash Pt: 146 °C(295 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006478 CAS Number: 2226-96-2 EINECS: 218-760-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...