Hydantoin 99%
Supplier: Thermo Scientific Chemicals
A12486-0E
A12486-22
A12486-36
CAAAA12486-0EEA
616.2
CAD
CAAAA12486-0E
CAAAA12486-22
CAAAA12486-36
Hydantoin 99%
Hydantoin
Purity: 99%
CAS No.: 461-72-3
Molecular Formula: C3H4N2O2
Formula Weight: 100.08
Melting Point: 221-223°
Beilstein Reference: 110598
Merck Reference: 14,4761
MDL No.: MFCD00005259
Formula:
C₃H₄N₂O₂ Melting Pt: 221…223 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005259 CAS Number: 461-72-3 EINECS: 207-313-3 Merck Index: 14,4761 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...