2-Mercaptobenzoxazole 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Benzo[d]oxazole-2-thiol

A12346-14 A12346-22
CAAAA12346-14EA 79.95 CAD
CAAAA12346-14 CAAAA12346-22
2-Mercaptobenzoxazole 98+%
2-Mercaptobenzoxazole

Purity: 98+% CAS No.: 2382-96-9 Molecular Formula: C7H5NOS Formula Weight: 151.19 Melting Point: 189-194° Beilstein Reference: 119483

Formula: C₇H₅NOS
MW: 151.19 g/mol
Melting Pt: 189…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00005769
CAS Number: 2382-96-9
EINECS: 219-191-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR