N-(Benzyloxycarbonyloxy)succinimide 98%
Supplier: Thermo Scientific Chemicals
A12153-14
A12153-22
CAAAA12153-06EA
34.57
CAD
CAAAA12153-06
CAAAA12153-14
CAAAA12153-22
N-(Benzyloxycarbonyloxy)succinimide 98%
N-(Benzyloxycarbonyloxy)succinimide
Purity: 99%
CAS No.: 13139-17-8
Molecular Formula: C12H11NO5
Formula Weight: 249.23
Storage Temperature: 2°C to 8°C
Melting Point: 77-82°
Beilstein Reference: 1387927
MDL No.: MFCD00005513
Formula:
C₁₂H₁₁NO₅ Melting Pt: 77…82 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005513 CAS Number: 13139-17-8 EINECS: 236-075-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...