Phosphonoacetic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

A12117-09 A12117-14 A12117-22
CAAAAA12117-09EA 145.83 CAD
CAAAAA12117-09 CAAAA12117-14 CAAAA12117-22
Phosphonoacetic acid 98+%
Phosphonoacetic acid

Purity: 98+% CAS No.: 4408-78-0 Molecular Formula: C2H5O5P Formula Weight: 140.04 Odor: Odorless Melting Point: 139-146° Beilstein Reference: 1764355 UN No.: 3261 MDL No.: MFCD00004311

Formula: C₂H₅O₅P
MW: 140.04 g/mol
Melting Pt: 139…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00004311
CAS Number: 4408-78-0
EINECS: 224-558-1
UN: 2923
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR