1-Phenyl-3-pyrazolidone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12089-0B
A12089-30
CAAAA12089-0BEA
663.6
CAD
CAAAA12089-0B
CAAAA12089-30
1-Phenyl-3-pyrazolidone 97%
1-Phenyl-3-pyrazolidone
Purity: 97%
CAS No.: 92-43-3
Molecular Formula: C9H10N2O
Formula Weight: 162.19
Melting Point: 120-123°
Beilstein Reference: 131856
Merck Reference: 14,7309
UN No.: 2811
MDL No.: MFCD00003094
Formula:
C₉H₁₀N₂O Melting Pt: 120…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003094 CAS Number: 92-43-3 EINECS: 202-155-1 UN: 2811 ADR: "6.1",III Merck Index: 14,7309 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...