Cacodylic acid (dimethylarsinic acid) 98%

Supplier: Thermo Scientific Chemicals
Danger

A12075-09 A12075-88
CAAAA12075-09EA 399.74 CAD
CAAAA12075-09 CAAAA12075-88
Cacodylic acid (dimethylarsinic acid) 98%
Cacodylic acid (Dimethylarsinic acid)

Purity: 98% CAS No.: 75-60-5 Molecular Formula: (CH3)2AsO2H Formula Weight: 138.00 Melting Point: 200° Beilstein Reference: 1736965 Merck Reference: 14,1604 UN No.: 1572 MDL No.: MFCD00002095

Formula: (CH₃)₂As(O)OH
Melting Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00002095
CAS Number: 75-60-5
EINECS: 200-883-4
UN: 1572
ADR: 6.1,II
Merck Index: 14,1604

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR