Dichloro(diphenyl)silane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Diphenyldichlorosilane, Dichlorodiphenylsilane
A12051-22
A12051-36
CAAAA12051-22EA
70.31
CAD
CAAAA12051-22
CAAAA12051-36
Dichloro(diphenyl)silane 97%
Dichloro(diphenyl)silane
Purity: 97%
CAS No.: 80-10-4
Molecular Formula: C12H10Cl2Si
Formula Weight: 253.20
Flash Point: 157°(314°F)
Refractive Index: 1.5790
Odor: Pungent
Melting Point: -22°
Boiling Point: 303-304°
Beilstein Reference: 609882
UN No.: 1769
MDL No.: MFCD00000489
Formula:
C₁₂H₁₀Cl₂Si MW: 253.2 g/mol Boiling Pt: 303…304 °C Melting Pt: –22 °C Density: 1.208 Flash Pt: 157 °C(314 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000489 CAS Number: 80-10-4 EINECS: 201-251-0 UN: 1769 ADR: "8",II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...