Methyl-2-methyl-3-furoate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-methyl-3-furoate

A11979-14 A11979-22
CAAAA11979-14EA 143.94 CAD
CAAAA11979-14 CAAAA11979-22
Methyl-2-methyl-3-furoate 97%
Methyl 2-methyl-3-furoate

Purity: 97% CAS No.: 6141-58-8 Molecular Formula: C7H8O3 Formula Weight: 140.14 Flash Point: 63°(145°F) Refractive Index: 1.4745 Boiling Point: 71-72°/20mm Beilstein Reference: 1342040 MDL No.: MFCD00003249

Formula: C₇H₈O₃
Boiling Pt: 71…72 °C/20 mm
Density: 1,114
Flash Pt: 63 °C(145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003249
CAS Number: 6141-58-8
EINECS: 228-132-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR